siRNA Details
| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037551.1 | - | 4 | 868 | ||
| AK084527.1 | - | 3 | 457 | |||
| AK169431.1 | - | 3 | 460 | |||
| BC008519.1 | - | 3 | 462 | 3592726 | ||
| BC085502.1 | - | 3 | 264 | 5720681 | ||
| L06039.1 | - | 3 | 347 | |||

Pecam1
Genomic Map