siRNA Details
| Gene Symbol: Peg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008817.2 | NP_032843.2 | 9 | 2643 | ||
| GenBank mRNA | AB003040.1 | - | 9 | 2643 | ||
| AF038939.1 | - | 6 | 2294 | |||
| AK044602.1 | - | 10 | 2707 | |||
| AK122238.1 | - | 9 | 2622 | |||
| AK141217.1 | - | 9 | 2644 | |||
| AK163845.1 | - | 9 | 2646 | |||
| BC048778.1 | - | 4 | 2020 | 6308481 | ||
| BC072661.1 | - | 9 | 2631 | 30543809 | ||
| BC085183.1 | - | 9 | 2628 | 30663888 | ||
| BC089344.1 | - | 1 | 1723 | |||

Peg3
Genomic Map