siRNA Details
| Gene Symbol: Peg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008817.2 | NP_032843.2 | 9 | 3303 | ||
| GenBank mRNA | AB003040.1 | - | 9 | 3303 | ||
| AK122238.1 | - | 9 | 3282 | |||
| AK141217.1 | - | 9 | 3304 | |||
| BC048778.1 | - | 4 | 2680 | 6308481 | ||
| BC072661.1 | - | 9, 10 | 3291 | 30543809 | ||
| BC085183.1 | - | 9 | 3288 | 30663888 | ||
| BC089344.1 | - | 1 | 2383 | |||

Peg3
Genomic Map