siRNA Details
| Gene Symbol: Sdr39u1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047709.1 | - | 730 | |||
| Gene Symbol: Penk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002927.2 | NP_001002927.1 | 1 | 385 | ||
| GenBank mRNA | AK161272.1 | - | 2 | 241 | ||
| BC049766.1 | - | 1 | 385 | 6774387 | ||
| M55181.1 | - | 1 | 379 | |||

Sdr39u1
Genomic Map