siRNA Details
| Gene Symbol: Pepd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008820.2 | NP_032846.2 | 2 | 198 | ||
| GenBank mRNA | AK010581.1 | - | 2 | 179 | ||
| AK145843.1 | - | 2 | 229 | |||
| AK168384.1 | - | 2 | 188 | |||
| BC086644.1 | - | 2 | 172 | 6807359 | ||
| BC131945.1 | - | 2 | 200 | 40130222 | ||
| BC131973.1 | - | 2 | 200 | 40130250 | ||
| D82983.1 | - | 2 | 185 | |||
| U51014.1 | - | 2 | 221 | |||

Pepd
Genomic Map