siRNA Details
| Gene Symbol: Per1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159367.1 | NP_001152839.1 | 8 | 1204 | ||
| NM_011065.4 | NP_035195.2 | 8 | 1219 | |||
| GenBank mRNA | AB002108.1 | - | 7 | 1033 | ||
| AF022992.1 | - | 8 | 1258 | |||
| AK081813.1 | - | 8 | 1217 | |||
| AK148202.1 | - | 8 | 1211 | |||
| AK154900.1 | - | 8 | 1200 | |||
| AK172958.1 | - | 8 | 1204 | |||
| BC039768.1 | - | 7 | 1104 | 4163009 | ||
| BC091645.1 | - | 8 | 1206 | 30355259 | ||

Per1
Genomic Map