siRNA Details
| Gene Symbol: Per3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289877.1 | NP_001276806.1 | 8 | 1380 | ||
| NM_001289878.1 | NP_001276807.1 | 9 | 1429 | |||
| NM_011067.3 | NP_035197.2 | 8 | 1380 | |||
| GenBank mRNA | AB013605.1 | - | 8 | 1187 | ||
| AF050182.1 | - | 8 | 1304 | |||
| AJ007377.1 | - | 8 | 1187 | |||
| AK036345.1 | - | 8 | 1297 | |||
| AK084610.1 | - | 1 | 2348 | |||
| AK166514.1 | - | 8 | 1381 | |||
| BC141032.1 | - | 8 | 978 | 9055644 | ||
| BC145178.1 | - | 9 | 1027 | |||

Per3
Genomic Map