siRNA Details
| Gene Symbol: Pex16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145122.2 | NP_660104.2 | 8 | 1071 | ||
| GenBank mRNA | AK020743.1 | - | 8 | 781 | ||
| AK143073.1 | - | 8 | 1073 | |||
| AK162716.1 | - | 8 | 771 | |||
| BC010822.1 | - | 8 | 812 | 4218547 | ||

Pex16
Genomic Map