siRNA Details
| Gene Symbol: Cfp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008823.3 | NP_032849.2 | 7, 8 | 1184 | ||
| GenBank mRNA | AK150212.1 | - | 7, 8 | 1145 | ||
| AK152782.1 | - | 7, 8 | 1135 | |||
| AK154306.1 | - | 7, 8 | 1169 | |||
| AK159291.1 | - | 7, 8 | 1185 | |||
| AK159609.1 | - | 7, 8 | 1134 | |||
| AK159853.1 | - | 7, 8 | 1134 | |||
| AK171374.1 | - | 7, 8 | 1168 | |||
| BC132369.1 | - | 7, 8 | 1168 | 40130646 | ||
| BC138306.1 | - | 7, 8 | 1168 | 8861326 | ||
| X12905.1 | - | 6, 7 | 1033 | |||

Cfp
Genomic Map