siRNA Details
| Gene Symbol: Pgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031190.2 | NP_112467.2 | 1 | 294 | ||
| GenBank mRNA | AK015016.1 | - | 1 | 285 | ||
| AK133436.1 | - | 1 | 283 | |||
| BC052343.1 | - | 1 | 258 | 6773764 | ||
| BC061054.1 | - | 1 | 285 | 6774063 | ||
| M18654.1 | - | 1 | 240 | |||

Pgk2
Genomic Map