siRNA Details
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008830.2 | NP_032856.2 | 8 | 976 | ||
GenBank mRNA | AK050345.1 | - | 7 | 929 | ||
BC150687.1 | - | 8 | 846 | 9087598 | ||
J03398.1 | - | 8 | 976 |
