siRNA Details
Gene Symbol: Piga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011081.2 | NP_035211.2 | 2 | 616 | ||
GenBank mRNA | AK032283.1 | - | 2 | 587 | ||
AK039802.1 | - | 2 | 616 | |||
AK161026.1 | - | 2 | 586 | |||
BC035538.1 | - | 1 | 277 | |||
BC138755.1 | - | 1 | 574 | 8861777 | ||
BC138758.1 | - | 1 | 574 | 8861780 | ||
D26047.1 | - | 2 | 585 | |||
S78188.1 | - | 2 | 622 |
