siRNA Details
| Gene Symbol: Pikfyve | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011086.2 | NP_035216.2 | 13, 14 | 1717 | ||
| GenBank mRNA | AF102777.1 | - | 13, 14 | 1697 | ||
| AK173077.1 | - | 13, 14 | 1787 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pikfyve| Gene Symbol: Pikfyve | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011086.2 | NP_035216.2 | 13, 14 | 1717 | ||
| GenBank mRNA | AF102777.1 | - | 13, 14 | 1697 | ||
| AK173077.1 | - | 13, 14 | 1787 | |||