siRNA Details
| Gene Symbol: Pikfyve | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011086.2 | NP_035216.2 | 20 | 3381 | ||
| GenBank mRNA | AF102777.1 | - | 20 | 3361 | ||
| AK017186.1 | - | 1 | 52 | |||
| AK173077.1 | - | 20 | 3451 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pikfyve| Gene Symbol: Pikfyve | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011086.2 | NP_035216.2 | 20 | 3381 | ||
| GenBank mRNA | AF102777.1 | - | 20 | 3361 | ||
| AK017186.1 | - | 1 | 52 | |||
| AK173077.1 | - | 20 | 3451 | |||