siRNA Details
| Gene Symbol: Prkaca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277898.1 | NP_001264827.1 | 10 | 1175 | ||
| NM_008854.5 | NP_032880.1 | 10 | 1218 | |||
| GenBank mRNA | BC003238.1 | - | 10 | 1208 | 3497908 | |
| BC054834.1 | - | 10 | 1203 | 6439034 | ||
| CB248596.1 | - | 2 | 115 | |||
| CD807077.1 | - | 6 | 622 | |||
| M12303.1 | - | 10 | 1195 | |||

Prkaca
Genomic Map