siRNA Details
| Gene Symbol: Pkp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019645.2 | NP_062619.1 | 1, 2 | 211 | ||
| GenBank mRNA | AK036856.1 | - | 1, 2 | 395 | ||
| AK079429.1 | - | 1, 2 | 395 | |||
| BC139041.1 | - | 1, 2 | 306 | 8862063 | ||
| BC139042.1 | - | 1, 2 | 307 | 8862064 | ||
| Y07941.1 | - | 1, 2 | 211 | |||

Pkp1
Genomic Map