siRNA Details
| Gene Symbol: Pla2r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008867.2 | NP_032893.1 | 7 | 1339 | ||
| GenBank mRNA | AK076366.1 | - | 7 | 1320 | ||
| AK143853.1 | - | 7 | 1318 | |||
| BC048780.1 | - | 7 | 1309 | |||
| BC141355.1 | - | 7 | 1220 | 9055967 | ||
| BC141356.1 | - | 7 | 1220 | 9055968 | ||
| D30779.1 | - | 7 | 1292 | |||

Pla2r1
Genomic Map