siRNA Details
| Gene Symbol: Plat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008872.2 | NP_032898.2 | 12 | 1429 | ||
| GenBank mRNA | AK135857.1 | - | 12 | 1420 | ||
| AK135965.1 | - | 12 | 1421 | |||
| AK141665.1 | - | 12 | 1418 | |||
| AK159620.1 | - | 12 | 1421 | |||
| AK166639.1 | - | 12 | 1419 | |||
| BC011256.1 | - | 12 | 1426 | 4038404 | ||
| BC057967.1 | - | 12 | 1409 | 4979664 | ||
| BC061508.1 | - | 12 | 1410 | 5352018 | ||
| J03520.1 | - | 12 | 1412 | |||

Plat
Genomic Map