siRNA Details
| Gene Symbol: Plat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008872.2 | NP_032898.2 | 14 | 1742 | ||
| GenBank mRNA | AK135857.1 | - | 14 | 1733 | ||
| AK135965.1 | - | 14 | 1734 | |||
| AK141665.1 | - | 14 | 1731 | |||
| AK159620.1 | - | 14 | 1734 | |||
| AK166639.1 | - | 14 | 1732 | |||
| BC011256.1 | - | 14 | 1739 | 4038404 | ||
| BC057967.1 | - | 14 | 1722 | 4979664 | ||
| BC061508.1 | - | 14 | 1723 | 5352018 | ||
| J03520.1 | - | 14 | 1725 | |||

Plat
Genomic Map