siRNA Details
| Gene Symbol: Plat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008872.2 | NP_032898.2 | 13 | 1538 | ||
| GenBank mRNA | AK135857.1 | - | 13 | 1529 | ||
| AK135965.1 | - | 13 | 1530 | |||
| AK159620.1 | - | 13 | 1530 | |||
| AK166639.1 | - | 13 | 1528 | |||
| BC011256.1 | - | 13 | 1535 | 4038404 | ||
| BC057967.1 | - | 13 | 1518 | 4979664 | ||
| BC061508.1 | - | 13 | 1519 | 5352018 | ||
| J03520.1 | - | 13 | 1521 | |||

Plat
Genomic Map