siRNA Details
| Gene Symbol: Plcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145830.1 | NP_001139302.1 | 21 | 2624 | ||
| NM_019677.2 | NP_062651.2 | 21 | 2624 | |||
| GenBank mRNA | AF498249.1 | - | 21 | 2255 | ||
| AF498250.1 | - | 21 | 2255 | |||
| AK129166.1 | - | 13 | 1489 | |||
| AK141164.1 | - | 21 | 2572 | |||
| BC058710.1 | - | 21 | 2591 | 6842016 | ||
| U85712.1 | - | 21 | 2258 | |||
| U85713.1 | - | 21 | 2258 | |||
| U85714.1 | - | 21 | 2258 | |||

Plcb1
Genomic Map