siRNA Details
| Gene Symbol: Plcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145830.1 | NP_001139302.1 | 4 | 665 | ||
| NM_019677.2 | NP_062651.2 | 4 | 665 | |||
| GenBank mRNA | AF022801.1 | - | 3 | 213 | ||
| AF498249.1 | - | 4 | 296 | |||
| AF498250.1 | - | 4 | 296 | |||
| AK141164.1 | - | 4 | 612 | |||
| AK141464.1 | - | 4 | 601 | |||
| AK143153.1 | - | 4 | 666 | |||
| BC058710.1 | - | 4 | 632 | 6842016 | ||
| U85712.1 | - | 4 | 299 | |||
| U85713.1 | - | 4 | 299 | |||
| U85714.1 | - | 4 | 299 | |||

Plcb1
Genomic Map