siRNA Details
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164056.1 | NP_001157528.1 | 20 | 2459 | ||
NM_008875.4 | NP_032901.2 | 19 | 2404 | |||
GenBank mRNA | AK136111.1 | - | 14 | 1642 | ||
BC068144.1 | - | 19 | 2261 | 30532470 | ||
U87868.1 | - | 20 | 2442 |
