siRNA Details
| Gene Symbol: Pld3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011116.1 | NP_035246.1 | 7 | 798 | ||
| GenBank mRNA | AF026124.1 | - | 7 | 798 | ||
| AK153508.1 | - | 5 | 487 | |||
| AK209515.1 | - | 1 | 36 | |||
| BC068170.1 | - | 7 | 795 | |||
| BC076586.1 | - | 7 | 862 | 30604186 | ||

Pld3
Genomic Map