siRNA Details
| Gene Symbol: Prl7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011120.2 | NP_035250.1 | 5 | 453 | ||
| GenBank mRNA | AK005421.1 | - | 5 | 450 | ||
| AK005557.1 | - | 5 | 453 | |||
| AK019461.1 | - | 5 | 453 | |||
| BC099470.1 | - | 5 | 442 | 30787403 | ||
| X02594.1 | - | 5 | 453 | |||

Prl7d1
Genomic Map