siRNA Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008877.3 | NP_032903.3 | 5 | 574 | ||
| GenBank mRNA | AK040840.1 | - | 5 | 525 | ||
| AK132254.1 | - | 5 | 543 | |||
| AY424276.1 | - | 5 | 542 | |||
| BC014773.1 | - | 5 | 575 | 4193957 | ||
| BC057186.1 | - | 5 | 549 | 5097123 | ||
| J04766.1 | - | 5 | 539 | |||

Plg
Genomic Map