siRNA Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008877.3 | NP_032903.3 | 15 | 1887 | ||
| GenBank mRNA | AK132254.1 | - | 15 | 1856 | ||
| BC014773.1 | - | 15 | 1888 | 4193957 | ||
| BC057186.1 | - | 15 | 1862 | 5097123 | ||
| J04766.1 | - | 15 | 1852 | |||

Plg
Genomic Map