siRNA Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008877.3 | NP_032903.3 | 15, 16 | 1936 | ||
| GenBank mRNA | AK132254.1 | - | 15, 16 | 1905 | ||
| BC014773.1 | - | 15, 16 | 1937 | 4193957 | ||
| BC057186.1 | - | 15, 16 | 1911 | 5097123 | ||
| J04766.1 | - | 15, 16 | 1901 | |||

Plg
Genomic Map