siRNA Details
Gene Symbol: Plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011121.3 | NP_035251.3 | 8 | 1463 | ||
GenBank mRNA | AK134045.1 | - | 8 | 1404 | ||
AK148050.1 | - | 8 | 1377 | |||
AK148076.1 | - | 8 | 1454 | |||
AK164027.1 | - | 8 | 1433 | |||
BC006880.1 | - | 8 | 1415 | 3596381 | ||
L19558.1 | - | 8 | 1470 | |||
U01063.1 | - | 8 | 1384 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
