siRNA Details
Gene Symbol: Plod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011122.3 | NP_035252.1 | 5, 6 | 680 | ||
GenBank mRNA | AF046782.1 | - | 5, 6 | 608 | ||
AK131632.1 | - | 5, 6 | 630 | |||
AK137480.1 | - | 5, 6 | 630 | |||
AK157729.1 | - | 5, 6 | 630 | |||
AK160822.1 | - | 5, 6 | 647 | |||
BC006599.1 | - | 5, 6 | 605 | 3965301 |
