siRNA Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011122.3 | NP_035252.1 | 4 | 441 | ||
| GenBank mRNA | AF046782.1 | - | 4 | 369 | ||
| AK131632.1 | - | 4 | 390 | |||
| AK137480.1 | - | 4 | 391 | |||
| AK157729.1 | - | 4 | 391 | |||
| AK160822.1 | - | 4 | 408 | |||
| BC006599.1 | - | 4 | 366 | 3965301 | ||

Plod1
Genomic Map