siRNA Details
| Gene Symbol: Plxna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008882.2 | NP_032908.2 | 17 | 3835 | ||
| GenBank mRNA | AK122289.1 | - | 16 | 2099 | ||
| BC068155.1 | - | 17 | 3617 | 30362726 | ||
| D86949.1 | - | 17 | 3741 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxna2| Gene Symbol: Plxna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008882.2 | NP_032908.2 | 17 | 3835 | ||
| GenBank mRNA | AK122289.1 | - | 16 | 2099 | ||
| BC068155.1 | - | 17 | 3617 | 30362726 | ||
| D86949.1 | - | 17 | 3741 | |||