siRNA Details
| Gene Symbol: Pml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008884.5 | NP_032910.3 | 8 | 2154 | ||
| NM_178087.4 | NP_835188.2 | 9 | 2292 | |||
| GenBank mRNA | AK028044.1 | - | 9 | 2294 | ||
| AK032799.1 | - | 7 | 1172 | |||
| AK137538.1 | - | 9 | 2235 | |||
| AK156279.1 | - | 9 | 2446 | |||
| BC020990.1 | - | 9 | 2099 | 4188386 | ||
| KJ650238.1 | - | 9 | 2196 | |||
| U33626.2 | - | 8 | 2051 | |||

Pml
Genomic Map