siRNA Details
| Gene Symbol: Pnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008891.2 | NP_032917.2 | 9 | 1303 | ||
| GenBank mRNA | AK028219.1 | - | 9 | 1244 | ||
| AK030978.1 | - | 9 | 1232 | |||
| AK050669.1 | - | 9 | 1249 | |||
| AK157587.1 | - | 9 | 1247 | |||
| AK160616.1 | - | 9 | 1238 | |||
| AK161109.1 | - | 9 | 1244 | |||
| BC132571.1 | - | 9 | 1237 | 40130848 | ||
| BC138014.1 | - | 9 | 1237 | 8861033 | ||
| Y08701.1 | - | 9 | 1234 | |||

Pnn
Genomic Map