siRNA Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284392.1 | NP_001271321.1 | 6 | 704 | ||
| NM_001284394.1 | NP_001271323.1 | 7 | 1001 | |||
| NM_001284398.1 | NP_001271327.1 | 6 | 688 | |||
| NM_011129.2 | NP_035259.1 | 7 | 1001 | |||
| GenBank mRNA | AB078010.1 | - | 6 | 599 | ||
| AK133933.1 | - | 6 | 656 | |||
| AK135143.1 | - | 7 | 1002 | |||
| BC055101.1 | - | 7 | 1001 | 6591016 | ||
| X61452.1 | - | 7 | 931 | |||

Sept4
Genomic Map