siRNA Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284394.1 | NP_001271323.1 | 2 | 369 | ||
| NM_011129.2 | NP_035259.1 | 2 | 369 | |||
| GenBank mRNA | AK039859.1 | - | 3 | 379 | ||
| AK135143.1 | - | 2 | 370 | |||
| AK137299.1 | - | 2 | 288 | |||
| BC055101.1 | - | 2 | 369 | 6591016 | ||
| BU562316.1 | - | 3 | 271 | |||
| X61452.1 | - | 2 | 299 | |||

Sept4
Genomic Map