siRNA Details
| Gene Symbol: Pon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011134.3 | NP_035264.2 | 6 | 801 | ||
| GenBank mRNA | BC012706.1 | - | 6 | 674 | 4158951 | |
| U32684.1 | - | 6 | 683 | |||
| U72636.1 | - | 6 | 683 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pon1| Gene Symbol: Pon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011134.3 | NP_035264.2 | 6 | 801 | ||
| GenBank mRNA | BC012706.1 | - | 6 | 674 | 4158951 | |
| U32684.1 | - | 6 | 683 | |||
| U72636.1 | - | 6 | 683 | |||