siRNA Details
| Gene Symbol: Endou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168693.1 | NP_001162164.1 | 7 | 955 | ||
| NM_008902.3 | NP_032928.1 | 6 | 954 | |||
| GenBank mRNA | AK132616.1 | - | 7 | 896 | ||
| AK169457.1 | - | 5 | 1045 | |||
| M95545.1 | - | 6 | 954 | |||

Endou
Genomic Map