siRNA Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113418.1 | NP_001106889.1 | 7 | 1125 | ||
| NM_011144.6 | NP_035274.2 | 7 | 1363 | |||
| GenBank mRNA | AK035676.1 | - | 7 | 1364 | ||
| AK081709.1 | - | 7 | 1126 | |||
| AK149460.1 | - | 7 | 1062 | |||
| BC016892.1 | - | 7 | 1356 | 4222384 | ||
| X57638.1 | - | 7 | 1084 | |||

Ppara
Genomic Map