siRNA Details
| Gene Symbol: Ppfibp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163557.1 | NP_001157029.1 | 11, 12 | 1181 | ||
| NM_008905.2 | NP_032931.2 | 11, 12 | 1417 | |||
| GenBank mRNA | AK012152.1 | - | 8, 9 | 840 | ||
| BC055935.1 | - | 1, 2 | 51 | |||
| BC100346.1 | - | 11, 12 | 1181 | 30936010 | ||
| U79024.1 | - | 11, 12 | 1202 | |||

Ppfibp2
Genomic Map