siRNA Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008911.2 | NP_032937.1 | 10, 11 | 1423 | ||
| GenBank mRNA | AK142632.1 | - | 10, 11 | 1375 | ||
| AK218241.1 | - | 4, 5 | 360 | |||
| BC002047.1 | - | 10, 11 | 1378 | 3484079 | ||
| D45185.1 | - | 10, 11 | 1149 | |||
| U25114.1 | - | 10, 11 | 1398 | |||

Ppox
Genomic Map