siRNA Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008911.2 | NP_032937.1 | 2, 3 | 406 | ||
| GenBank mRNA | AK134341.1 | - | 346 | |||
| AK142632.1 | - | 2, 3 | 358 | |||
| BC002047.1 | - | 2, 3 | 361 | 3484079 | ||
| D45185.1 | - | 2, 3 | 132 | |||
| U25114.1 | - | 2, 3 | 381 | |||

Ppox
Genomic Map