siRNA Details
| Gene Symbol: Ppp3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293622.1 | NP_001280551.1 | 2 | 903 | ||
| NM_008913.5 | NP_032939.1 | 2 | 903 | |||
| GenBank mRNA | AK146387.1 | - | 2 | 847 | ||
| AK150393.1 | - | 2 | 262 | |||
| AK166287.1 | - | 2 | 678 | |||
| AK209023.1 | - | 1 | 44 | |||
| BC096652.1 | - | 2 | 145 | |||
| BC138612.1 | - | 2 | 176 | 8861632 | ||
| J05479.1 | - | 2 | 200 | |||

Ppp3ca
Genomic Map