siRNA Details
| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015002.1 | - | 4 | 665 | ||
| AK133393.1 | - | 4 | 686 | |||
| BC046617.1 | - | 4 | 491 | |||
| BC141079.1 | - | 4 | 506 | 9055691 | ||
| BC145347.1 | - | 3 | 381 | |||
| M81475.1 | - | 4 | 659 | |||

Ppp3cc
Genomic Map