siRNA Details
| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133393.1 | - | 14 | 1649 | ||
| BC046617.1 | - | 15 | 1481 | |||
| BC141079.1 | - | 14 | 1469 | 9055691 | ||
| BC145347.1 | - | 13 | 1344 | |||
| M81475.1 | - | 14 | 1622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp3cc| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133393.1 | - | 14 | 1649 | ||
| BC046617.1 | - | 15 | 1481 | |||
| BC141079.1 | - | 14 | 1469 | 9055691 | ||
| BC145347.1 | - | 13 | 1344 | |||
| M81475.1 | - | 14 | 1622 | |||