siRNA Details
| Gene Symbol: Prep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011156.2 | NP_035286.1 | 8 | 1036 | ||
| GenBank mRNA | AB007631.1 | - | 8 | 919 | ||
| AK053816.1 | - | 8 | 1047 | |||
| AK076257.1 | - | 8 | 1035 | |||
| AK133875.1 | - | 8 | 1035 | |||
| AK141851.1 | - | 8 | 1045 | |||
| AK161528.1 | - | 8 | 1035 | |||
| AK163787.1 | - | 8 | 1048 | |||
| AK170570.1 | - | 8 | 1045 | |||
| BC012869.1 | - | 8 | 1036 | 3967219 | ||
| BC050830.1 | - | 8 | 1029 | 6519113 | ||

Prep
Genomic Map