siRNA Details
| Gene Symbol: Mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011161.5 | NP_035291.4 | 8, 9 | 764 | ||
| GenBank mRNA | AF135185.1 | - | 8, 9 | 744 | ||
| AK144846.1 | - | 8, 9 | 737 | |||
| AK161562.1 | - | 8, 9 | 716 | |||
| BC057211.1 | - | 8, 9 | 700 | |||
| BC064737.1 | - | 8, 9 | 715 | 30112690 | ||
| BC092526.1 | - | 8, 9 | 764 | 6593321 | ||

Mapk11
Genomic Map