siRNA Details
| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008929.3 | NP_032955.2 | 9 | 1136 | ||
| GenBank mRNA | AK004033.1 | - | 4 | 437 | ||
| AK146097.1 | - | 9 | 1100 | |||
| AK148270.1 | - | 9 | 1093 | |||
| AK188318.1 | - | 3 | 211 | |||
| BC013766.1 | - | 9 | 1081 | 2646147 | ||
| U28423.1 | - | 9 | 1079 | |||

Dnajc3
Genomic Map