siRNA Details
| Gene Symbol: Prlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011169.5 | NP_035299.4 | 10 | 2454 | ||
| GenBank mRNA | AK054551.1 | - | 9 | 2965 | ||
| AK077980.1 | - | 1 | 95 | |||
| BC005555.1 | - | 10 | 2037 | 3586308 | ||
| BC006652.1 | - | 10 | 1928 | 3500076 | ||
| D10214.1 | - | 8 | 1729 | |||
| L13593.1 | - | 8 | 1764 | |||
| L14811.1 | - | 8 | 1706 | |||
| X73372.1 | - | 8 | 1706 | |||

Prlr
Genomic Map