siRNA Details
| Gene Symbol: Prodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011172.2 | NP_035302.2 | 2, 3 | 508 | ||
| GenBank mRNA | AF120279.1 | - | 2, 3 | 465 | ||
| AK144068.1 | - | 2, 3 | 478 | |||
| BB611875.1 | - | 2, 3 | 507 | |||
| BC008548.1 | - | 2, 3 | 332 | |||
| BC125327.1 | - | 2, 3 | 311 | 40129842 | ||
| U80020.1 | - | 2, 3 | 457 | |||

Prodh
Genomic Map